missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1-chloro-2-nitrobenzene 98.0+%, TCI America™
$74.56 - $218.10
Chemical Identifiers
| CAS | 16588-24-2 |
|---|---|
| Molecular Formula | C6H3BrClNO2 |
| Molecular Weight (g/mol) | 236.45 |
| MDL Number | MFCD05863216 |
| InChI Key | DJRYWPGOQTUJMQ-UHFFFAOYSA-N |
| Synonym | 5-Bromo-2-chloronitrobenzene |
| PubChem CID | 817974 |
| IUPAC Name | 4-bromo-1-chloro-2-nitrobenzene |
| SMILES | [O-][N+](=O)C1=C(Cl)C=CC(Br)=C1 |
Chemical Identifiers
| 16588-24-2 | |
| 236.45 | |
| DJRYWPGOQTUJMQ-UHFFFAOYSA-N | |
| 817974 | |
| [O-][N+](=O)C1=C(Cl)C=CC(Br)=C1 |
| C6H3BrClNO2 | |
| MFCD05863216 | |
| 5-Bromo-2-chloronitrobenzene | |
| 4-bromo-1-chloro-2-nitrobenzene |
Specifications
| 16588-24-2 | |
| Yellow | |
| MFCD05863216 | |
| 5-Bromo-2-chloronitrobenzene | |
| [O-][N+](=O)C1=C(Cl)C=CC(Br)=C1 | |
| 236.45 | |
| 236.45 | |
| Crystalline Powder |
| 72°C | |
| C6H3BrClNO2 | |
| 5 g | |
| DJRYWPGOQTUJMQ-UHFFFAOYSA-N | |
| 4-bromo-1-chloro-2-nitrobenzene | |
| 817974 | |
| ≥98.0% (GC) | |
| 4-Bromo-1-chloro-2-nitrobenzene |
Safety and Handling
TSCA : No