missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butyne 97.0+%, TCI America™

Catalog No. B32425G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32425G Supplier TCI America Supplier No. B32425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38771-21-0
Molecular Formula C4H5Br
Molecular Weight (g/mol) 132.988
InChI Key XLYOGWXIKVUXCL-UHFFFAOYSA-N
Synonym 4-bromo-1-butyne, 1-bromo-3-butyne, 1-butyne, 4-bromo, 4-bromobutyne, 3-butynyl bromide, 1-bromo-3-butynol, 4-bromo-but-1-yne, acmc-1afup, 1-butyne, 4-bromo-6ci,9ci
PubChem CID 11073464
IUPAC Name 4-bromobut-1-yne
SMILES C#CCCBr

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
UN Number 1993
Formula Weight 132.99
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-butyne