accessibility menu, dialog, popup

UserName

CAS RN 38771-21-0

IUPAC Name: 4-bromobut-1-yne
Synonyms: 4-bromobut-1-yne
Molecular Weight (g/mol): 132.99
Molecular Formula: C4H5Br
InChi Key: XLYOGWXIKVUXCL-UHFFFAOYSA-N
SMILES: BrCCC#C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)