missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butene 97.0+%, TCI America™

Catalog No. p-7118743
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B092010G 10 g
B092025G 25 g
B0920250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B092010G Supplier TCI America Supplier No. B092010G
Only null left

Chemical Identifiers

CAS 5162-44-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.004
MDL Number MFCD00000258
InChI Key DMAYBPBPEUFIHJ-UHFFFAOYSA-N
Synonym 4-bromo-1-butene, 3-butenyl bromide, 1-butene, 4-bromo, 4-bromo-1-buten, 4-bromo-but-1-ene, homoallyl bromide, 1-bromo-3-butene, 4-bromobutene, 4-bromo-1 butene, sjphabijuh@
PubChem CID 21241
IUPAC Name 4-bromobut-1-ene
SMILES C=CCCBr

Specifications

Color Yellow
Boiling Point 99°C
Quantity 10 g
UN Number 1993
Formula Weight 135.00
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-butene