missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butene, 98+%

Catalog No. p-73000
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173550050 5 mL Glass bottle
AC173550250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173550050 Supplier Thermo Scientific Chemicals Supplier No. 173550050
Only null left

CAS: 5162-44-7 | C4H7Br | 135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5162-44-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135
MDL Number MFCD00000258
InChI Key DMAYBPBPEUFIHJ-UHFFFAOYSA-N
Synonym 4-bromo-1-butene, 3-butenyl bromide, 1-butene, 4-bromo, 4-bromo-1-buten, 4-bromo-but-1-ene, homoallyl bromide, 1-bromo-3-butene, 4-bromobutene, 4-bromo-1 butene, sjphabijuh@
PubChem CID 21241
IUPAC Name 4-bromobut-1-ene
SMILES C=CCCBr

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Brown to Colorless
Density 1.3300g/mL
Boiling Point 98.0°C to 100.0°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula BrCH2CH2CH=CH2
Refractive Index 1.4595 to 1.4635
Quantity 5 mL
Beilstein 01, I, 84
Specific Gravity 1.33
Formula Weight 135
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-butene, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
May cause an allergic skin reaction.
Highly flammable liquid and va
Precautionary Statement GHS P Statement
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear protective gloves/pr
EINECSNumber 225-937-4
TSCA TSCA

RUO – Research Use Only