missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,3-benzodioxole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC01710DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC01710DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

6698-13-1
201.019
VZPMQHSDFWAZHP-UHFFFAOYSA-N
2776189
C1OC2=C(O1)C(=CC=C2)Br
C7H5BrO2
MFCD02681890
4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide
4-bromo-1,3-benzodioxole

Specifications

4-Bromo-1, 3-benzodioxole
96.5
97%
C7H5BrO2
MFCD02681890
VZPMQHSDFWAZHP-UHFFFAOYSA-N
4-bromo-1,3-benzodioxole
2776189
97%
6698-13-1
100.0
Amber Glass Bottle
1g
4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide
C1OC2=C(O1)C(=CC=C2)Br
201.019
201.02
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents