missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,3-benzodioxole, 97%, Thermo Scientific™

Catalog No. p-2701352
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC01710CB 250mg Amber Glass Bottle
CC01710DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC01710CB Supplier Thermo Scientific Chemicals Supplier No. CC01710CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6698-13-1
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD02681890
InChI Key VZPMQHSDFWAZHP-UHFFFAOYSA-N
Synonym 4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide
PubChem CID 2776189
IUPAC Name 4-bromo-1,3-benzodioxole
SMILES C1OC2=C(O1)C(=CC=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 201.02
Percent Purity 97%
Chemical Name or Material 4-Bromo-1, 3-benzodioxole
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.