missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-difluorobenzene, 98+%

Catalog No. p-7081789
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1193406 5 g
AAA1193414 25 g
AAA1193422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1193406 Supplier Thermo Scientific Chemicals Supplier No. A1193406
Only null left

CAS: 348-61-8 | C6H3BrF2 | 192.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-61-8
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00000304
InChI Key YMQPKONILWWJQG-UHFFFAOYSA-N
Synonym 1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide
PubChem CID 67675
IUPAC Name 4-bromo-1,2-difluorobenzene
SMILES FC1=CC=C(Br)C=C1F

Specifications

Melting Point -4°C
Density 1.707
Boiling Point 150°C to 151°C
Flash Point 33°C (91°F)
Refractive Index 1.505
Quantity 5 g
UN Number UN1993
Beilstein 1934811
Formula Weight 192.99
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-1,2-difluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-481-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only