missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,1,7,7-tetraethyl-1,2,3,5,6,7-hexahydro-3,3,5,5-tetramethyl-s-indacene 95.0+%, TCI America™

Catalog No. p-7123502
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43791G 1 g
B43795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43791G Supplier TCI America Supplier No. B43791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1142818-90-3
Molecular Formula C24H37Br
Molecular Weight (g/mol) 405.46
MDL Number MFCD28100820
InChI Key DTVNULIVVOTHAI-UHFFFAOYSA-N
Synonym EMind-Br
PubChem CID 91972079
IUPAC Name 4-bromo-1,1,7,7-tetraethyl-3,3,5,5-tetramethyl-1,2,3,5,6,7-hexahydro-s-indacene
SMILES CCC1(CC)CC(C)(C)C2=C(Br)C3=C(C=C12)C(CC)(CC)CC3(C)C

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 405.46
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1,1,7,7-tetraethyl-1,2,3,5,6,7-hexahydro-3,3,5,5-tetramethyl-s-indacene