missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 4-Borono-L-phenylalanine, 97%

Chemical Identifiers

CAS 76410-58-7
Molecular Formula C9H12BNO4
Molecular Weight (g/mol) 209.01
MDL Number MFCD01075172
InChI Key NFIVJOSXJDORSP-QMMMGPOBSA-N
Synonym 4-borono-l-phenylalanine, s-2-amino-3-4-boronophenyl propanoic acid, 4-boronophenylalanine, p-boronophenylalanine, para-boronophenylalanine, 10b-bpa, unii-jsm63lv5md, para-borono-l-phenylalanine, l-phe b oh 2, l-4-boronophenylalanine
PubChem CID 150315
IUPAC Name (2S)-2-amino-3-(4-boronophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(C=C1)B(O)O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC358931000
SDS View Documents
Thermo Scientific™
358931000
100mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC358932500
SDS View Documents
Thermo Scientific™
358932500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

76410-58-7
209.01
NFIVJOSXJDORSP-QMMMGPOBSA-N
150315
N[C@@H](CC1=CC=C(C=C1)B(O)O)C(O)=O
C9H12BNO4
MFCD01075172
4-borono-l-phenylalanine, s-2-amino-3-4-boronophenyl propanoic acid, 4-boronophenylalanine, p-boronophenylalanine, para-boronophenylalanine, 10b-bpa, unii-jsm63lv5md, para-borono-l-phenylalanine, l-phe b oh 2, l-4-boronophenylalanine
(2S)-2-amino-3-(4-boronophenyl)propanoic acid

Specifications

76410-58-7
100.0
96% min. (HPLC)
C9H12BNO4
100mg
NFIVJOSXJDORSP-QMMMGPOBSA-N
(2S)-2-amino-3-(4-boronophenyl)propanoic acid
150315
97%
96.0
Authentic
Glass bottle
MFCD01075172
4-borono-l-phenylalanine, s-2-amino-3-4-boronophenyl propanoic acid, 4-boronophenylalanine, p-boronophenylalanine, para-boronophenylalanine, 10b-bpa, unii-jsm63lv5md, para-borono-l-phenylalanine, l-phe b oh 2, l-4-boronophenylalanine
N[C@@H](CC1=CC=C(C=C1)B(O)O)C(O)=O
209.01
209.01
4-Borono-L-phenylalanine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only