CAS RN 76410-58-7
IUPAC Name:
(2S)-2-amino-3-[4-(dihydroxyboranyl)phenyl]propanoic acid
Synonyms:
boronophenylalanine
Molecular Weight (g/mol):
209.01
Molecular Formula:
C9H12BNO4
InChi Key:
NFIVJOSXJDORSP-QMMMGPOBSA-N
SMILES:
N[C@@H](CC1=CC=C(C=C1)B(O)O)C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 4-Borono-L-phenylalanine, 97%
CAS: 76410-58-7 Molecular Formula: C9H12BNO4 Molecular Weight (g/mol): 209.01 MDL Number: MFCD01075172 InChI Key: NFIVJOSXJDORSP-QMMMGPOBSA-N Synonym: 4-borono-l-phenylalanine,s-2-amino-3-4-boronophenyl propanoic acid,4-boronophenylalanine,p-boronophenylalanine,para-boronophenylalanine,10b-bpa,unii-jsm63lv5md,para-borono-l-phenylalanine,l-phe b oh 2,l-4-boronophenylalanine PubChem CID: 150315 IUPAC Name: (2S)-2-amino-3-(4-boronophenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(C=C1)B(O)O)C(O)=O