missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Borono-L-phenylalanine, 97%

Catalog No. AC358931000
Change view
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC358931000 100 mg Glass bottle
AC358932500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC358931000 Supplier Thermo Scientific Chemicals Supplier No. 358931000
Only null left

CAS: 76410-58-7 | C9H12BNO4 | 209.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76410-58-7
Molecular Formula C9H12BNO4
Molecular Weight (g/mol) 209.01
MDL Number MFCD01075172
InChI Key NFIVJOSXJDORSP-QMMMGPOBSA-N
Synonym 4-borono-l-phenylalanine, s-2-amino-3-4-boronophenyl propanoic acid, 4-boronophenylalanine, p-boronophenylalanine, para-boronophenylalanine, 10b-bpa, unii-jsm63lv5md, para-borono-l-phenylalanine, l-phe b oh 2, l-4-boronophenylalanine
PubChem CID 150315
IUPAC Name (2S)-2-amino-3-(4-boronophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(C=C1)B(O)O)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 283.0°C to 293.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 209.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Borono-L-phenylalanine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only