Learn More
4-(Boc-aminomethyl)piperidine, 97%
CAS: 135632-53-0 | C11H22N2O2 | 214.309 g/mol
$121.29 - $469.58
Chemical Identifiers
| CAS | 135632-53-0 |
|---|---|
| Molecular Formula | C11H22N2O2 |
| Molecular Weight (g/mol) | 214.309 |
| MDL Number | MFCD01631214 |
| InChI Key | VHYXAWLOJGIJPC-UHFFFAOYSA-N |
| Synonym | tert-butyl piperidin-4-ylmethyl carbamate, 4-boc-aminomethyl piperidine, 4-tert-butoxycarbonylaminomethyl piperidine, 4-n-boc-aminomethyl piperidine, 4-boc-aminomethyl-piperidine, tert-butyl n-piperidin-4-ylmethyl carbamate, tert-butyl piperidin-4-ylmethylcarbamate, boc-4-aminomethyl piperidine |
| PubChem CID | 723429 |
| IUPAC Name | tert-butyl N-(piperidin-4-ylmethyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NCC1CCNCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5272503
|
Thermo Scientific Chemicals
H5272503 |
1 g |
Each for $121.29
|
|
|||||
|
AAH5272506
|
Thermo Scientific Chemicals
H5272506 |
5 g |
Each for $469.58
|
|
|||||
Description
4-(Boc-aminomethyl)piperidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Boc-aminomethyl)piperidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in DMSO.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
| 135632-53-0 | |
| 214.309 | |
| VHYXAWLOJGIJPC-UHFFFAOYSA-N | |
| 723429 | |
| CC(C)(C)OC(=O)NCC1CCNCC1 |
| C11H22N2O2 | |
| MFCD01631214 | |
| tert-butyl piperidin-4-ylmethyl carbamate, 4-boc-aminomethyl piperidine, 4-tert-butoxycarbonylaminomethyl piperidine, 4-n-boc-aminomethyl piperidine, 4-boc-aminomethyl-piperidine, tert-butyl n-piperidin-4-ylmethyl carbamate, tert-butyl piperidin-4-ylmethylcarbamate, boc-4-aminomethyl piperidine | |
| tert-butyl N-(piperidin-4-ylmethyl)carbamate |
Specifications
| 106°C to 108°C | |
| C11H22N2O2 | |
| UN3259 | |
| Soluble in DMSO. | |
| CC(C)(C)OC(=O)NCC1CCNCC1 | |
| 1 g | |
| 723429 | |
| 97% |
| 135632-53-0 | |
| MFCD01631214 | |
| tert-butyl piperidin-4-ylmethyl carbamate, 4-boc-aminomethyl piperidine, 4-tert-butoxycarbonylaminomethyl piperidine, 4-n-boc-aminomethyl piperidine, 4-boc-aminomethyl-piperidine, tert-butyl n-piperidin-4-ylmethyl carbamate, tert-butyl piperidin-4-ylmethylcarbamate, boc-4-aminomethyl piperidine | |
| VHYXAWLOJGIJPC-UHFFFAOYSA-N | |
| tert-butyl N-(piperidin-4-ylmethyl)carbamate | |
| 214.309 | |
| 214.31 | |
| 4-(Boc-aminomethyl)piperidine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only