accessibility menu, dialog, popup

UserName

CAS RN 135632-53-0

IUPAC Name: tert-butyl N-[(pyridin-4-yl)methyl]carbamate
Synonyms: tert-butyl N-(pyridin-4-ylmethyl)carbamate
Molecular Weight (g/mol): 208.26
Molecular Formula: C11H16N2O2
InChi Key: KXCFJDZVQLRYSI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=NC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)