missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-aminomethyl)piperidine, 97%

Catalog No. AAH5272503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5272503 1 g
AAH5272506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5272503 Supplier Thermo Scientific Chemicals Supplier No. H5272503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 135632-53-0 | C11H22N2O2 | 214.309 g/mol

4-(Boc-aminomethyl)piperidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-aminomethyl)piperidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 135632-53-0
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD01631214
InChI Key VHYXAWLOJGIJPC-UHFFFAOYSA-N
Synonym tert-butyl piperidin-4-ylmethyl carbamate, 4-boc-aminomethyl piperidine, 4-tert-butoxycarbonylaminomethyl piperidine, 4-n-boc-aminomethyl piperidine, 4-boc-aminomethyl-piperidine, tert-butyl n-piperidin-4-ylmethyl carbamate, tert-butyl piperidin-4-ylmethylcarbamate, boc-4-aminomethyl piperidine
PubChem CID 723429
IUPAC Name tert-butyl N-(piperidin-4-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1CCNCC1

Specifications

Melting Point 106°C to 108°C
UN Number UN3259
Solubility Information Soluble in DMSO.
Quantity 1 g
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material 4-(Boc-aminomethyl)piperidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only