missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-aminomethyl)benzoic acid, 97%

Catalog No. p-7057061
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6639006 5 g
AAH6639014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6639006 Supplier Thermo Scientific Chemicals Supplier No. H6639006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33233-67-9 | C13H16NO4 | 250.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33233-67-9
Molecular Formula C13H16NO4
Molecular Weight (g/mol) 250.28
MDL Number MFCD00228182
InChI Key LNKHBRDWRIIROP-UHFFFAOYSA-M
Synonym 4-tert-butoxycarbonylamino methyl benzoic acid, 4-tert-butoxycarbonyl amino methyl benzoic acid, 4-tert-butoxy carbonyl amino methyl benzoic acid, boc-4-aminomethyl-benzoic acid, 4-boc-aminomethyl-benzoic acid, 4-tert-butoxycarbonylaminomethyl benzoic acid, 4-n-tert-butyloxycarbonyl amino methyl benzoic acid, benzoic acid, 4-1,1-dimethylethoxy carbonyl amino methyl
PubChem CID 853605
IUPAC Name 4-({[(tert-butoxy)carbonyl]amino}methyl)benzoate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C([O-])=O

Specifications

Melting Point 164°C to 168°C
Color White
Beilstein 2854286
Quantity 5 g
Formula Weight 251.28
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 4-(Boc-aminomethyl)benzoic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only