Learn More
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6294803
Description
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid generally used in the synthesis of organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid generally used in the synthesis of organic compounds.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
| 4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester | |
| C18H27BFNO4 | |
| 4-(Boc-aminomethyl)-3-fluorophenylboronic acid pinacol ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CNC(=O)OC(C)(C)C)F | |
| 351.225 | |
| 351.22 |
| 1351501-44-4 | |
| 1 g | |
| SAUVAJYHMCIQNE-UHFFFAOYSA-N | |
| tert-butyl N-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate | |
| 66980292 | |
| 96% |
Chemical Identifiers
| 1351501-44-4 | |
| 351.225 | |
| 4-(Boc-aminomethyl)-3-fluorophenylboronic acid pinacol ester | |
| tert-butyl N-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate |
| C18H27BFNO4 | |
| SAUVAJYHMCIQNE-UHFFFAOYSA-N | |
| 66980292 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CNC(=O)OC(C)(C)C)F |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only