missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)tetrahydropyran-4-carboxylic acid, 95%

Catalog No. p-7030047
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27578MD 250 mg
AAH2757803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27578MD Supplier Thermo Scientific Chemicals Supplier No. H27578MD
Only null left

CAS: 172843-97-9 | C11H19NO5 | 245.275 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water. (15 g/L) (25°C)

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 172843-97-9
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.275
MDL Number MFCD02683136
InChI Key SPPDKPRJPFTBEV-UHFFFAOYSA-N
Synonym 4-boc-amino tetrahydropyran-4-carboxylic acid, 4-n-boc-amino-4-carboxytetrahydropyran, 4-tert-butoxycarbonyl amino tetrahydro-2h-pyran-4-carboxylic acid, 4-tert-butoxycarbonyl amino oxane-4-carboxylic acid, 4-tert-butoxy carbonyl amino oxane-4-carboxylic acid, 4-tert-butoxycarbamoyl tetrahydropyran-4-carboxylic acid, 4-tert-butoxycarbonylamino tetrahydro-2h-pyran-4-carboxylic acid, 4-tert-butoxy carbonylamino-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
PubChem CID 1268219
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]oxane-4-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCOCC1)C(=O)O

Specifications

Melting Point 157°C (decomposition)
Quantity 250 mg
Solubility Information Soluble in water. (15g/L) (25°C)
Formula Weight 245.27
Percent Purity 95%
Chemical Name or Material 4-(Boc-amino)tetrahydropyran-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only