accessibility menu, dialog, popup

UserName

CAS RN 172843-97-9

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}oxane-4-carboxylate
Synonyms: 4-[(tert-butoxycarbonyl)amino]oxane-4-carboxylate
Molecular Weight (g/mol): 244.27
Molecular Formula: C11H18NO5
InChi Key: SPPDKPRJPFTBEV-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)NC1(CCOCC1)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)