missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)benzeneboronic acid pinacol ester, 97%

Catalog No. p-7047709
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1965303 1 g
AAL1965306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1965303 Supplier Thermo Scientific Chemicals Supplier No. L1965303

CAS: 330793-01-6 | C17H26BNO4 | 319.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 330793-01-6
Molecular Formula C17H26BNO4
Molecular Weight (g/mol) 319.21
MDL Number MFCD02179439
InChI Key HSJNIOYPTSKQBD-UHFFFAOYSA-N
Synonym 4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
PubChem CID 2734617
IUPAC Name tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 169°C to 170°C
Quantity 1 g
Formula Weight 319.21
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)benzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only