missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)-2-chlorobenzoic acid, 97%

Catalog No. AAH5278603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5278603 1 g
AAH5278606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5278603 Supplier Thermo Scientific Chemicals Supplier No. H5278603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 232275-73-9 | C12H14ClNO4 | 271.697 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 232275-73-9
Molecular Formula C12H14ClNO4
Molecular Weight (g/mol) 271.697
MDL Number MFCD02682164
InChI Key HDYBNYDKCZJPPX-UHFFFAOYSA-N
Synonym n-boc-4-amino-2-chlorobenzoic acid, 4-tert-butoxycarbonyl amino-2-chlorobenzoic acid, 2-chloro-4-boc-amino-benzoic acid, 4-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-4-boc-amino benzoic acid, 4-tert-butoxycarbonylamino-2-chloro-benzoic acid, 4-tert-butoxycarbonylamino-2-chlorobenzoic acid, benzoic acid,2-chloro-4-1,1-dimethylethoxy carbonyl amino
PubChem CID 17750449
IUPAC Name 2-chloro-4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC(=C(C=C1)C(=O)O)Cl

Specifications

Quantity 1 g
Formula Weight 271.7
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)-2-chlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only