missing translation for 'onlineSavingsMsg'
Learn More

4-Boc-1-(6-methyl-2-pyridyl)piperazine, 97%, Thermo Scientific™

$124.62 - $124.62

Chemical Identifiers

CAS 127188-33-4
Molecular Formula C15H23N3O2
Molecular Weight (g/mol) 277.368
MDL Number MFCD07369771
InChI Key QEDVZUFNZJJSJL-UHFFFAOYSA-N
Synonym 4-boc-1-6-methyl-2-pyridyl piperazine, tert-butyl 4-6-methylpyridin-2-yl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-6-methyl-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1c1la, 4-boc-1-6-methylpyridin-2-yl piperazine, tert-butyl-4-6-methylpyridin-2-yl piperazine-1-carboxylate
PubChem CID 16217938
IUPAC Name tert-butyl 4-(6-methylpyridin-2-yl)piperazine-1-carboxylate
SMILES CC1=NC(=CC=C1)N2CCN(CC2)C(=O)OC(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5584103
View Documents
Thermo Scientific Chemicals
H5584103
1 g
Each for $124.62
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

127188-33-4
277.368
QEDVZUFNZJJSJL-UHFFFAOYSA-N
16217938
CC1=NC(=CC=C1)N2CCN(CC2)C(=O)OC(C)(C)C
C15H23N3O2
MFCD07369771
4-boc-1-6-methyl-2-pyridyl piperazine, tert-butyl 4-6-methylpyridin-2-yl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-6-methyl-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1c1la, 4-boc-1-6-methylpyridin-2-yl piperazine, tert-butyl-4-6-methylpyridin-2-yl piperazine-1-carboxylate
tert-butyl 4-(6-methylpyridin-2-yl)piperazine-1-carboxylate

Specifications

127188-33-4
White
MFCD07369771
UN2811
QEDVZUFNZJJSJL-UHFFFAOYSA-N
tert-butyl 4-(6-methylpyridin-2-yl)piperazine-1-carboxylate
16217938
97%
4-Boc-1-(6-methyl-2-pyridyl)piperazine
80°C to 84°C
C15H23N3O2
1 g
4-boc-1-6-methyl-2-pyridyl piperazine, tert-butyl 4-6-methylpyridin-2-yl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-6-methyl-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1c1la, 4-boc-1-6-methylpyridin-2-yl piperazine, tert-butyl-4-6-methylpyridin-2-yl piperazine-1-carboxylate
CC1=NC(=CC=C1)N2CCN(CC2)C(=O)OC(C)(C)C
277.368
277.37
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'