accessibility menu, dialog, popup

UserName

CAS RN 127188-33-4

IUPAC Name: tert-butyl 4-(6-methylpyridin-2-yl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(6-methylpyridin-2-yl)piperazine-1-carboxylate
Molecular Weight (g/mol): 277.37
Molecular Formula: C15H23N3O2
InChi Key: QEDVZUFNZJJSJL-UHFFFAOYSA-N
SMILES: CC1=NC(=CC=C1)N1CCN(CC1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results