missing translation for 'onlineSavingsMsg'
Learn More

4-[Bis(4-iodophenyl)amino]benzaldehyde 95.0+%, TCI America™

Catalog No. B4543200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4543200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4543200MG Supplier TCI America Supplier No. B4543200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 808758-81-8
Molecular Formula C19H13I2NO
Molecular Weight (g/mol) 525.128
InChI Key HIUBJHKIRVFTOM-UHFFFAOYSA-N
PubChem CID 58130137
IUPAC Name 4-(4-iodo-N-(4-iodophenyl)anilino)benzaldehyde
SMILES C1=CC(=CC=C1C=O)N(C2=CC=C(C=C2)I)C3=CC=C(C=C3)I

Specifications

Melting Point 140°C
Quantity 200 mg
Formula Weight 525.13
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[Bis(4-iodophenyl)amino]benzaldehyde