accessibility menu, dialog, popup

UserName

CAS RN 808758-81-8

IUPAC Name: 4-[bis(4-iodophenyl)amino]benzaldehyde
Synonyms: 4-[bis(4-iodophenyl)amino]benzaldehyde
Molecular Weight (g/mol): 525.13
Molecular Formula: C19H13I2NO
InChi Key: HIUBJHKIRVFTOM-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)N(C1=CC=C(I)C=C1)C1=CC=C(C=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results