missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Biphenylmethanol, 98+%
CAS: 3597-91-9 | C13H12O | 184.238 g/mol
$88.99 - $1551.70
Chemical Identifiers
| CAS | 3597-91-9 |
|---|---|
| Molecular Formula | C13H12O |
| Molecular Weight (g/mol) | 184.238 |
| MDL Number | MFCD00004660 |
| InChI Key | AXCHZLOJGKSWLV-UHFFFAOYSA-N |
| Synonym | 4-biphenylmethanol, 4-phenylphenyl methanol, biphenyl-4-ylmethanol, 4-phenylbenzyl alcohol, p-phenylbenzyl alcohol, 4-hydroxymethylbiphenyl, 1,1'-biphenyl-4-ylmethanol, biphenyl-4-methanol, 4-hydroxymethyl biphenyl, 4hmb |
| PubChem CID | 19186 |
| IUPAC Name | (4-phenylphenyl)methanol |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1187609
|
Thermo Scientific Chemicals
A1187609 |
10 g |
Each for $88.99
|
|
|||||
|
AAA1187618
|
Thermo Scientific Chemicals
A1187618 |
50 g |
Each for $293.04
|
|
|||||
|
AAA1187630
|
Thermo Scientific Chemicals
A1187630 |
250 g |
Each for $1,551.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3597-91-9 | |
| 184.238 | |
| AXCHZLOJGKSWLV-UHFFFAOYSA-N | |
| 19186 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)CO |
| C13H12O | |
| MFCD00004660 | |
| 4-biphenylmethanol, 4-phenylphenyl methanol, biphenyl-4-ylmethanol, 4-phenylbenzyl alcohol, p-phenylbenzyl alcohol, 4-hydroxymethylbiphenyl, 1,1'-biphenyl-4-ylmethanol, biphenyl-4-methanol, 4-hydroxymethyl biphenyl, 4hmb | |
| (4-phenylphenyl)methanol |
Specifications
| 3597-91-9 | |
| 184°C (11 mmHg) | |
| MFCD00004660 | |
| 1937761 | |
| AXCHZLOJGKSWLV-UHFFFAOYSA-N | |
| (4-phenylphenyl)methanol | |
| 19186 | |
| ≥98% |
| 98°C to 104°C | |
| C13H12O | |
| 10 g | |
| 4-biphenylmethanol, 4-phenylphenyl methanol, biphenyl-4-ylmethanol, 4-phenylbenzyl alcohol, p-phenylbenzyl alcohol, 4-hydroxymethylbiphenyl, 1,1'-biphenyl-4-ylmethanol, biphenyl-4-methanol, 4-hydroxymethyl biphenyl, 4hmb | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)CO | |
| 184.238 | |
| 184.24 | |
| 4-Biphenylmethanol |
Safety and Handling
EINECSNumber : 222-745-2
RTECSNumber : DV5425000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only