missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylmethanol, 98+%

Catalog No. AAA1187609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1187609 10 g
AAA1187618 50 g
AAA1187630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1187609 Supplier Thermo Scientific Chemicals Supplier No. A1187609

CAS: 3597-91-9 | C13H12O | 184.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3597-91-9
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00004660
InChI Key AXCHZLOJGKSWLV-UHFFFAOYSA-N
Synonym 4-biphenylmethanol, 4-phenylphenyl methanol, biphenyl-4-ylmethanol, 4-phenylbenzyl alcohol, p-phenylbenzyl alcohol, 4-hydroxymethylbiphenyl, 1,1'-biphenyl-4-ylmethanol, biphenyl-4-methanol, 4-hydroxymethyl biphenyl, 4hmb
PubChem CID 19186
IUPAC Name (4-phenylphenyl)methanol
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CO

Specifications

Melting Point 98°C to 104°C
Boiling Point 184°C (11 mmHg)
Quantity 10 g
Beilstein 1937761
Formula Weight 184.24
Percent Purity ≥98%
Chemical Name or Material 4-Biphenylmethanol
EINECSNumber 222-745-2
RTECSNumber DV5425000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only