missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylcarboxylic acid, 98%

Catalog No. AC106281000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106281000 100 g Plastic bottle
AC106280250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106281000 Supplier Thermo Scientific Chemicals Supplier No. 106281000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92-92-2 | C13H10O2 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-92-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002553
InChI Key NNJMFJSKMRYHSR-UHFFFAOYSA-N
Synonym 4-biphenylcarboxylic acid, biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, p-phenylbenzoic acid, 4-carboxybiphenyl, para phenyl benzoic acid, 4-diphenylcarboxylic acid, diphenyl-4-carboxylic acid, 4-carboxy-1,1'-biphenyl
PubChem CID 66724
IUPAC Name 4-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 223°C to 228°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5C6H4CO2H
Quantity 100 g
Beilstein 09, 671
Formula Weight 198.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenylcarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-203-1

RUO – Research Use Only