missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylacetic Acid 98.0+%, TCI America™

Catalog No. B127825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B127825G 25 g
B1278250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B127825G Supplier TCI America Supplier No. B127825G

Chemical Identifiers

CAS 5728-52-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00004351
InChI Key QRZAKQDHEVVFRX-UHFFFAOYSA-N
Synonym 4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
PubChem CID 3332
ChEBI CHEBI:31597
IUPAC Name 2-(4-phenylphenyl)acetic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 212.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenylacetic Acid
RTECSNumber DU8229050
TSCA No