accessibility menu, dialog, popup

UserName

OHO

CAS RN 5728-52-9

IUPAC Name: 2-{[1,1'-biphenyl]-4-yl}acetic acid
Synonyms: felbinac dolinac felbinacum felbinaco biphenyl-4-yl-acetaldehyde traxam 联苯乙酸 napageln
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: QRZAKQDHEVVFRX-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)