missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylacetic acid, 98%

CAS: 5728-52-9 | C14H12O2 | 212.248 g/mol

$137.50 - $137.50

Chemical Identifiers

CAS 5728-52-9
MDL Number MFCD00004351
InChI Key QRZAKQDHEVVFRX-UHFFFAOYSA-N
Synonym 4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
PubChem CID 3332
ChEBI CHEBI:31597
IUPAC Name 2-(4-phenylphenyl)acetic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC188150100
SDS View Documents
Thermo Scientific Chemicals
188150100
10 g Glass bottle
Each for $137.50
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5728-52-9
QRZAKQDHEVVFRX-UHFFFAOYSA-N
3332
2-(4-phenylphenyl)acetic acid
MFCD00004351
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
CHEBI:31597
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O

Specifications

5728-52-9
100.0
Beige
98%
C14H12O2
MFCD00004351
09, I, 284
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O
212.25
CHEBI:31597
98%
4-Biphenylacetic acid
97.5
156°C to 160°C
Authentic
Glass bottle
C6H5C6H4CH2CO2H
10 g
15, 3985
QRZAKQDHEVVFRX-UHFFFAOYSA-N
2-(4-phenylphenyl)acetic acid
3332
212.25
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 227-233-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only