missing translation for 'onlineSavingsMsg'
Learn More

4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine 98.0+%, TCI America™

Catalog No. B49531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49531G 1 g
B49535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49531G Supplier TCI America Supplier No. B49531G

Chemical Identifiers

CAS 1421599-34-9
Molecular Formula C28H19BrN2
Molecular Weight (g/mol) 463.38
MDL Number MFCD28975104
InChI Key VSCGHXGRCSQGIE-UHFFFAOYSA-N
PubChem CID 89992073
IUPAC Name 4-{[1,1'-biphenyl]-4-yl}-6-(4-bromophenyl)-2-phenylpyrimidine
SMILES BrC1=CC=C(C=C1)C1=NC(=NC(=C1)C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 1 g
Formula Weight 463.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine