accessibility menu, dialog, popup

UserName

NNBr

CAS RN 1421599-34-9

IUPAC Name: 4-{[1,1'-biphenyl]-4-yl}-6-(4-bromophenyl)-2-phenylpyrimidine
Synonyms: 4-{[1,1'-biphenyl]-4-yl}-6-(4-bromophenyl)-2-phenylpyrimidine
Molecular Weight (g/mol): 463.38
Molecular Formula: C28H19BrN2
InChi Key: VSCGHXGRCSQGIE-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=NC(=NC(=C1)C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results