missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzylphenol, Thermo Scientific™
$64.38 - $210.61
Chemical Identifiers
| CAS | 101-53-1 |
|---|---|
| Molecular Formula | C13H12O |
| Molecular Weight (g/mol) | 184.24 |
| MDL Number | 00002384 |
| InChI Key | HJSPWKGEPDZNLK-UHFFFAOYSA-N |
| IUPAC Name | 4-benzylphenol |
| SMILES | OC1=CC=C(CC2=CC=CC=C2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC121130010
|
Thermo Scientific Chemicals
121130010 |
1 g |
Each for $64.38
|
|
|||||
|
AC121130050
|
Thermo Scientific Chemicals
121130050 |
5 g |
Each for $210.61
|
|
|||||
Chemical Identifiers
| 101-53-1 | |
| 184.24 | |
| HJSPWKGEPDZNLK-UHFFFAOYSA-N | |
| OC1=CC=C(CC2=CC=CC=C2)C=C1 |
| C13H12O | |
| 00002384 | |
| 4-benzylphenol |
Specifications
| BENZYLPHENOL | |
| 80°C to 85°C | |
| Conforms | |
| C13H12O | |
| 00002384 | |
| 06-675 | |
| HJSPWKGEPDZNLK-UHFFFAOYSA-N | |
| 4-benzylphenol | |
| Crystalline Powder or Crystals |
| 101-53-1 | |
| White to beige to brown | |
| Glass Bottle | |
| C6H5CH2C6H4OH | |
| 1 g | |
| 15-1145 | |
| OC1=CC=C(CC2=CC=CC=C2)C=C1 | |
| 184.24 | |
| 4-Benzylphenol |
Safety and Handling
Exclamation mark
EINECSNumber : 202-950-3
RTECSNumber : SM8330500
Recommended Storage : Normal conditions