missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzylphenol, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 121130050
Specifications
| BENZYLPHENOL | |
| 101-53-1 | |
| White to beige to brown | |
| Glass Bottle | |
| C6H5CH2C6H4OH | |
| 5 g | |
| 15-1145 | |
| OC1=CC=C(CC2=CC=CC=C2)C=C1 | |
| 184.24 |
| 4-Benzylphenol | |
| 80°C to 85°C | |
| Conforms | |
| C13H12O | |
| 00002384 | |
| 06-675 | |
| HJSPWKGEPDZNLK-UHFFFAOYSA-N | |
| 4-benzylphenol | |
| Crystalline Powder or Crystals |
Chemical Identifiers
| 101-53-1 | |
| 184.24 | |
| HJSPWKGEPDZNLK-UHFFFAOYSA-N | |
| OC1=CC=C(CC2=CC=CC=C2)C=C1 |
| C13H12O | |
| 00002384 | |
| 4-benzylphenol |
Safety and Handling
Exclamation mark
EINECSNumber : 202-950-3
RTECSNumber : SM8330500
Recommended Storage : Normal conditions