missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylphenol 98.0+%, TCI America™

$150.98 - $1537.84

Chemical Identifiers

CAS 101-53-1
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00002384
InChI Key HJSPWKGEPDZNLK-UHFFFAOYSA-N
Synonym p-benzylphenol, fesiasept, 4-hydroxydiphenylmethane, 4-hydroxyditane, 4-phenylmethyl phenol, phenol, 4-phenylmethyl, p-hydroxydiphenyl methane, alpha-phenyl-p-cresol, p-hydroxydiphenylmethane, 4-benzyl-phenol
PubChem CID 7563
IUPAC Name 4-benzylphenol
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H023925G
View Documents
TCI America
H023925G
25 g
Each for $150.98
Only null left
 
H0239500G
View Documents
TCI America
H0239500G
500 g
Each for $1,537.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

101-53-1
184.238
HJSPWKGEPDZNLK-UHFFFAOYSA-N
7563
C1=CC=C(C=C1)CC2=CC=C(C=C2)O
C13H12O
MFCD00002384
p-benzylphenol, fesiasept, 4-hydroxydiphenylmethane, 4-hydroxyditane, 4-phenylmethyl phenol, phenol, 4-phenylmethyl, p-hydroxydiphenyl methane, alpha-phenyl-p-cresol, p-hydroxydiphenylmethane, 4-benzyl-phenol
4-benzylphenol

Specifications

101-53-1
White-Yellow
C13H12O
25 g
HJSPWKGEPDZNLK-UHFFFAOYSA-N
4-benzylphenol
7563
≥98.0% (GC)
4-Benzylphenol
83°C
330°C
MFCD00002384
p-benzylphenol, fesiasept, 4-hydroxydiphenylmethane, 4-hydroxyditane, 4-phenylmethyl phenol, phenol, 4-phenylmethyl, p-hydroxydiphenyl methane, alpha-phenyl-p-cresol, p-hydroxydiphenylmethane, 4-benzyl-phenol
C1=CC=C(C=C1)CC2=CC=C(C=C2)O
184.238
184.24
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0547

RTECSNumber : SM8330500

TSCA : Yes

SDS
Documents

Documents