missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylphenol 98.0+%, TCI America™

Catalog No. H023925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H023925G 25 g
H0239500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H023925G Supplier TCI America Supplier No. H023925G

Chemical Identifiers

CAS 101-53-1
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00002384
InChI Key HJSPWKGEPDZNLK-UHFFFAOYSA-N
Synonym p-benzylphenol, fesiasept, 4-hydroxydiphenylmethane, 4-hydroxyditane, 4-phenylmethyl phenol, phenol, 4-phenylmethyl, p-hydroxydiphenyl methane, alpha-phenyl-p-cresol, p-hydroxydiphenylmethane, 4-benzyl-phenol
PubChem CID 7563
IUPAC Name 4-benzylphenol
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)O

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 330°C
Quantity 25 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzylphenol
EINECSNumber (3)-0547
RTECSNumber SM8330500
TSCA Yes