missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenylmagnesium bromide, 0.8M solution in THF, AcroSeal™

Catalog No. AC436221000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436221000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436221000 Supplier Thermo Scientific Chemicals Supplier No. 436221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120186-59-6 | C13H11BrMgO | 287.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120186-59-6 , 109-99-9
Molecular Formula C13H11BrMgO
Molecular Weight (g/mol) 287.44
MDL Number MFCD09039130
InChI Key NMSSOXCJYITCNG-UHFFFAOYSA-M
Synonym 4-benzyloxyphenylmagnesium bromide solution, magnesium bromide 4-benzyloxy benzen-1-ide 1/1/1, magnesium 2+ ion 4-benzyloxy benzen-1-ide bromide, grignard reagent
PubChem CID 11098170
IUPAC Name magnesium;phenylmethoxybenzene;bromide
SMILES Br[Mg]C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

CAS Min % 19.48, 74.96
CAS Max % 25.04, 80.52
Color Colorless to Brown
Density 1.0330g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula C6H5CH2OC6H4MgBr
Quantity 100 mL
Specific Gravity 1.033
Formula Weight 287.43
Physical Form Liquid
Chemical Name or Material 4-Benzyloxyphenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only