missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AC394030050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC394030050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC394030050 Supplier Thermo Scientific Chemicals Supplier No. 394030050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52068-30-1 | Molecular Formula: C13H14N2O·HCl | Molecular Weight: 250.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52068-30-1
Molecular Formula C13H14N2O·HCl
Molecular Weight (g/mol) 250.73
MDL Number MFCD00137741
InChI Key OVNUPJXMCMTQCN-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylhydrazine hydrochloride, 4-benzyloxy phenyl hydrazine hydrochloride, 4-benzyloxyphenyl hydrazine hydrochloride, 4-benzyloxy phenyl hydrazine monohydrochloride, 4-benzyloxyphenyl hydrazine hcl, 4-benzyloxyphenylhydrazine hcl, 4-benzyloxyphenylhydrazinehydrochloride, 4-phenylmethoxyphenyl hydrazine hydrochloride
PubChem CID 3016725
IUPAC Name (4-phenylmethoxyphenyl)hydrazine;hydrochloride
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)NN.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 187°C to 189°C
Color Brown to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4NHNH2·HCl
Quantity 5 g
Formula Weight 250.73
Percent Purity 98%
Physical Form Fluffy Powder
Chemical Name or Material 4-Benzyloxyphenylhydrazine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 257-636-9

RUO – Research Use Only