missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenyl 4-Hydroxyphenyl Sulfone 98.0+%, TCI America™

Catalog No. B28585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28585G Supplier TCI America Supplier No. B28585G
Only null left

Chemical Identifiers

CAS 63134-33-8
Molecular Formula C19H16O4S
Molecular Weight (g/mol) 340.393
InChI Key UWPJWCBDMZHMTN-UHFFFAOYSA-N
PubChem CID 113063
IUPAC Name 4-(4-phenylmethoxyphenyl)sulfonylphenol
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)S(=O)(=O)C3=CC=C(C=C3)O

Specifications

Melting Point 170°C
Color White
Quantity 5 g
Formula Weight 340.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxyphenyl 4-Hydroxyphenyl Sulfone
EINECSNumber (3)-4366
TSCA Yes