missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7119194
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16655G 5 g
B166525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16655G Supplier TCI America Supplier No. B16655G

Chemical Identifiers

CAS 1486-51-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00016527
InChI Key AQSCHALQLXXKKC-UHFFFAOYSA-N
PubChem CID 73880
IUPAC Name 4-(benzyloxy)benzoic acid
SMILES OC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 190°C
Color White
Quantity 5 g
Synonym 4-benzyloxybenzoic acid, 4-benzyloxy benzoic acid, benzoic acid, 4-phenylmethoxy, p-benzyloxy benzoic acid, 4-phenylmethoxy benzoic acid, 4-benzyloxy-benzoic acid, 4-benzyl-oxy benzoic acid, benzoic acid,4-phenylmethoxy, asischem a95731
Formula Weight 228.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzoic Acid