missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzoic acid, 98%

Catalog No. p-7022800
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1571503 1 g
AAA1571506 5 g
AAA1571514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1571503 Supplier Thermo Scientific Chemicals Supplier No. A1571503
Only null left

CAS: 1486-51-7 | C14H12O3 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1486-51-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00016527
InChI Key AQSCHALQLXXKKC-UHFFFAOYSA-N
PubChem CID 73880
IUPAC Name 4-phenylmethoxybenzoic acid
SMILES OC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 187°C to 191°C
Quantity 1 g
Beilstein 2111640
Synonym 4-benzyloxybenzoic acid, 4-benzyloxy benzoic acid, benzoic acid, 4-phenylmethoxy, p-benzyloxy benzoic acid, 4-phenylmethoxy benzoic acid, 4-benzyloxy-benzoic acid, 4-benzyl-oxy benzoic acid, benzoic acid,4-phenylmethoxy, asischem a95731
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 4-Benzyloxybenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-066-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only