missing translation for 'onlineSavingsMsg'
Learn More

4'-(Benzyloxy)benzylidene-4-fluoroaniline 98.0+%, TCI America™

Catalog No. p-7123456
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B43015G 5 g
B430125G 25 g
2 options

Catalog No. B43015G

Supplier: TCI America B43015G

Only null left

Chemical Identifiers

CAS 70627-52-0
Molecular Formula C20H16FNO
Molecular Weight (g/mol) 305.35
MDL Number MFCD00017951
InChI Key IWNBEFDVKWCBFY-UHFFFAOYSA-N
PubChem CID 791783
IUPAC Name 1-[4-(benzyloxy)phenyl]-N-(4-fluorophenyl)methanimine
SMILES FC1=CC=C(C=C1)N=CC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 5 g
Formula Weight 305.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(Benzyloxy)benzylidene-4-fluoroaniline