missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-(Benzyloxy)benzylidene-4-fluoroaniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 136°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 305.35 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4′-(Benzyloxy)benzylidene-4-fluoroaniline |
Safety and Handling
| TSCA | No |