missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)benzyl Chloride 98.0+%, TCI America™

Catalog No. B51211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51211G 1 g
B51215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51211G Supplier TCI America Supplier No. B51211G
Only null left

Chemical Identifiers

CAS 836-42-0
Molecular Formula C14H13ClO
Molecular Weight (g/mol) 232.707
MDL Number MFCD00000916
InChI Key UYQPSKUPEXAQRJ-UHFFFAOYSA-N
Synonym Benzyl 4-(Chloromethyl)phenyl Ether, 1-(Benzyloxy)-4-(chloromethyl)benzene
PubChem CID 70042
IUPAC Name 1-(chloromethyl)-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)CCl

Specifications

Melting Point 79°C
Quantity 1 g
UN Number 3261
Formula Weight 232.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Benzyloxy)benzyl Chloride
TSCA No
Recommended Storage Refrigerator