missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog No. AAH2744306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2744306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2744306 Supplier Thermo Scientific Chemicals Supplier No. H2744306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33693-48-0
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00004640
InChI Key PDBXFVPMVYQICB-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxybenzyl alcohol, 4-benzyloxy-3-methoxyphenyl methanol, 4-benzyloxy-3-methoxybenzylalcohol, benzenemethanol, 3-methoxy-4-phenylmethoxy, acmc-20amos, o-benzylvanillyl alcohol, 4-o-benzyl vanillyl alcohol, vanillyl alcohol benzyl ether
PubChem CID 98832
IUPAC Name (3-methoxy-4-phenylmethoxyphenyl)methanol
SMILES COC1=C(C=CC(=C1)CO)OCC2=CC=CC=C2

Specifications

Melting Point 71°C to 73°C
Quantity 5 g
Formula Weight 244.3
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-3-methoxybenzyl alcohol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 251-638-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only