missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyloxy-3-methoxybenzaldehyde 98.0+%, TCI America™
$72.70 - $175.22
Chemical Identifiers
| CAS | 2426-87-1 |
|---|---|
| Molecular Formula | C15H14O3 |
| Molecular Weight (g/mol) | 242.27 |
| MDL Number | MFCD00003365 |
| InChI Key | JSHLOPGSDZTEGQ-UHFFFAOYSA-N |
| Synonym | 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde |
| PubChem CID | 75506 |
| IUPAC Name | 4-(benzyloxy)-3-methoxybenzaldehyde |
| SMILES | COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1 |
Chemical Identifiers
| 2426-87-1 | |
| 242.27 | |
| JSHLOPGSDZTEGQ-UHFFFAOYSA-N | |
| 75506 | |
| COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1 |
| C15H14O3 | |
| MFCD00003365 | |
| 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde | |
| 4-(benzyloxy)-3-methoxybenzaldehyde |
Specifications
| 2426-87-1 | |
| Yellow | |
| C15H14O3 | |
| 5 g | |
| JSHLOPGSDZTEGQ-UHFFFAOYSA-N | |
| 4-(benzyloxy)-3-methoxybenzaldehyde | |
| 75506 | |
| ≥98.0% (GC) | |
| 4-Benzyloxy-3-methoxybenzaldehyde |
| 63°C | |
| 185°C | |
| MFCD00003365 | |
| 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde | |
| COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1 | |
| 242.27 | |
| 242.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No