missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxybenzaldehyde 98.0+%, TCI America™

Catalog No. B35995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35995G 5 g
B359925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35995G Supplier TCI America Supplier No. B35995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2426-87-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00003365
InChI Key JSHLOPGSDZTEGQ-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde
PubChem CID 75506
IUPAC Name 4-(benzyloxy)-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 63°C
Color Yellow
Boiling Point 185°C
Quantity 5 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxy-3-methoxybenzaldehyde