missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxybenzaldehyde, 98%

Catalog No. AAA1471614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1471614 25 g
AAA1471622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1471614 Supplier Thermo Scientific Chemicals Supplier No. A1471614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2426-87-1 | C15H14O3 | 242.27 g/mol

4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N?-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N′-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.

Solubility
Soluble in chloroform, methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 2426-87-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00003365
InChI Key JSHLOPGSDZTEGQ-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde
PubChem CID 75506
IUPAC Name 3-methoxy-4-phenylmethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 61°C to 65°C
Boiling Point 213°C to 215°C (3.5 mmHg)
Quantity 25 g
Beilstein 1464258
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,methanol.
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-3-methoxybenzaldehyde
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 219-379-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only