missing translation for 'onlineSavingsMsg'
Learn More

4'-Benzyloxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog No. AAL0843006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0843006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0843006 Supplier Thermo Scientific Chemicals Supplier No. L0843006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1835-11-6 | Molecular Formula: C16H16O3 | Molecular Weight: 256.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1835-11-6
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00017326
InChI Key HRUAWSQBQLYDKH-UHFFFAOYSA-N
Synonym 1-4-benzyloxy-3-methoxyphenyl ethanone, 4'-benzyloxy-3'-methoxyacetophenone, 4-benzyloxy-3-methoxyacetophenone, acetophenone, 4'-benzyloxy-3'-methoxy, 1-3-methoxy-4-phenylmethoxy phenyl ethanone, 1-4-benzyloxy-3-methoxyphenyl ethan-1-one, 1-3-methoxy-4-phenylmethoxyphenyl ethanone, ethanone, 1-3-methoxy-4-phenylmethoxy phenyl
PubChem CID 99215
IUPAC Name 1-(3-methoxy-4-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OCC2=CC=CC=C2)OC

Specifications

Melting Point 85°C to 87°C
Quantity 5 g
Beilstein 1885776
Formula Weight 256.3
Percent Purity 98%
Chemical Name or Material 4'-Benzyloxy-3'-methoxyacetophenone
RTECSNumber AM5966000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only